Learn More
Acetaldoxime, 99%, mixture of syn and anti, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 102230250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Acetaldoxime | |
107-29-9 | |
100.0 | |
115.0°C | |
Authentic | |
Glass bottle | |
1.4250 to 1.4270 | |
MFCD00002124 | |
01, IV, 3121 | |
0.96 | |
acetaldoxime, z-acetaldehyde oxime, 1z-acetaldehyde oxime, acetaldehyde, oxime, 1z, 1z-ethanal oxime, nz-n-ethylidenehydroxylamine, acetaldehyde, oxime, acetaldehyde, oxime, z, z-acetaldoxime, cis-acetaldehyde oxime | |
FZENGILVLUJGJX-IHWYPQMZSA-N | |
(NZ)-N-ethylidenehydroxylamine | |
5324280 | |
59.06 |
99% | |
98.5 | |
0.9600g/mL | |
38°C | |
98.5% min. (GC) | |
C2H5NO | |
CH3CH=NOH | |
25g | |
01,3 | |
15, 43 | |
Solubility in water: soluble in water. Other solubilities: decomposes by aq. hcl,soluble in alcohol and ether | |
CC=NO | |
59.06 | |
CHEBI:50719 | |
99% |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Flammable solid.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 203-479-6
RTECSNumber : AB2975000
TSCA : TSCA
RUO â Research Use Only