Please login to your online account to display your discounted pricing

Acetaldehyde, SPEX CertiPrep™

$119.91 - $119.91

Chemical Identifiers

Chemical Name or Material Acetaldehyde
CAS 75-07-0
CAS Min % 0.1
CAS Max % 0.1
CAS 67-56-1
CAS Min % 99.9
CAS Max % 99.9
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S-125

 
spex certiprep llc
S125
2mL amber prescored glass ampule 1mL Each for $119.91
Description & Specifications

Specifications

Quantity 1mL
Packaging 2mL amber prescored glass ampule
DOT Information Nonhazardous
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Health Hazard 1 Highly flammable liquid and vapor. Toxic if inhaled. Causes damage to organs.
Recommended Storage Ambient
Solvent or Matrix Methanol-P&T
Analyte or Component Names Acetaldehyde
Chemical Name or Material Acetaldehyde
CAS Min % 0.1
CAS 67-56-1
CAS Max % 0.1
CAS Max % 99.9
CAS Min % 99.9
CAS 75-07-0

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Highly flammable liquid and vapor. Toxic if inhaled. Causes damage to organs.

Ambient