Learn More
Acetaldehyde ammonia trimer, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 148535000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Acetaldehyde ammonia trimer | |
58052-80-5 | |
100.0 | |
55°C | |
98% | |
C6H18N3 | |
500g | |
2,4,6-trimethyl-1,3,5-triazinane trihydrate, acetaldehyde ammonia trimer, hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate, 1,3,5-triazine, hexahydro-2,4,6-trimethyl-, trihydrate, acetaldehyde ammonia trimer trihydrate, 2,4,6-trimethyl-1,3,5-triazaperhydroine, hydrate, hydrate, hydrate, acetaldehydeammoniatrimer, acmc-1bm0n, acetaldehyde-ammonia trimer trihydrate, acetaldehyde ammonia trimer nt | |
MZSSRMMSFLVKPK-UHFFFAOYSA-Q | |
2,4,6-trimethyl-1,3,5-triazinane;trihydrate | |
2723814 | |
98% |
98% | |
97.5 | |
110.0°C | |
Authentic | |
Glass bottle | |
MFCD00149559 | |
26, I, 3 | |
Solubility in water: very soluble. Other solubilities: soluble in ethanol,chloroform | |
CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1 | |
132.23 | |
183.26 |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Warning
EINECSNumber : 200-868-2
RUO â Research Use Only