missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 148530250

 View more versions of this product

Catalog No. AC148530250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acetaldehyde ammonia trimer
58052-80-5
100.0
Beige
55°C
98%
C6H18N3
25 g
2,4,6-trimethyl-1,3,5-triazinane trihydrate, acetaldehyde ammonia trimer, hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate, 1,3,5-triazine, hexahydro-2,4,6-trimethyl-, trihydrate, acetaldehyde ammonia trimer trihydrate, 2,4,6-trimethyl-1,3,5-triazaperhydroine, hydrate, hydrate, hydrate, acetaldehydeammoniatrimer, acmc-1bm0n, acetaldehyde-ammonia trimer trihydrate, acetaldehyde ammonia trimer nt
MZSSRMMSFLVKPK-UHFFFAOYSA-Q
2,4,6-trimethyl-1,3,5-triazinane;trihydrate
2723814
98%
98%
97.5
95.0°C to 97.0°C
110.0°C
Authentic
Glass bottle
MFCD00149559
26, I, 3
Solubility in water: very soluble. Other solubilities: soluble in ethanol,chloroform
CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1
132.23
183.26
Crystalline Powder

Chemical Identifiers

58052-80-5
132.23
MZSSRMMSFLVKPK-UHFFFAOYSA-Q
2723814
C6H18N3
MFCD00149559
2,4,6-trimethyl-1,3,5-triazinane trihydrate, acetaldehyde ammonia trimer, hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate, 1,3,5-triazine, hexahydro-2,4,6-trimethyl-, trihydrate, acetaldehyde ammonia trimer trihydrate, 2,4,6-trimethyl-1,3,5-triazaperhydroine, hydrate, hydrate, hydrate, acetaldehydeammoniatrimer, acmc-1bm0n, acetaldehyde-ammonia trimer trihydrate, acetaldehyde ammonia trimer nt
CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning

EINECSNumber : 200-868-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only