Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetaldehyde ammonia trimer, 98%

Chemical Identifiers

Chemical Name or Material Acetaldehyde ammonia trimer
CAS 58052-80-5
Molecular Formula C6H15N3·3H2O
Formula Weight 183.25
MDL Number MFCD00149559
UN Number UN1841
Beilstein 3910586
Merck Index 14,40
Synonym Hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1226518

 
alfa aesar™
L1226518
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 183.25
MDL Number MFCD00149559
Chemical Name or Material Acetaldehyde ammonia trimer
Assay 98%
Molecular Formula C6H15N3·3H2O
CAS 58052-80-5
Merck Index 14,40
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 110°C (dec.)
Quantity 50g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN1841
Beilstein 3910586
Synonym Hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate
Health Hazard 1 Warning
Melting Point 93° to 96°C
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ACETALDEHYDE AMMONIA

Applications
Acetaldehyde ammonia trimer is used as scavengers for sulfhydryl compounds in natural gas.

Solubility
Soluble in water.

Notes
Desiccate at 4°C. Store away from air, moisture and oxidizing agents. Store under dry inert gas. Protect from humidity and water.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.