Learn More
Acetaldehyde, 99.5%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 149510100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acetaldehyde | |
75-07-0 | |
100.0 | |
Colorless | |
21.0°C | |
Authentic | |
Pungent | |
C2H4O | |
CH3CHO | |
10 mL | |
01,3 | |
15, 40 | |
Solubility in water: > 500g/L (20°C). Other solubilities: soluble in ethanol and acetone | |
CC=O | |
44.04 | |
CHEBI:15343 | |
99.5% | |
Volatile Liquid |
Extra Pure | |
99.4 | |
-123.0°C | |
0.7850g/mL | |
−27°C | |
99.4% min. (GC) | |
Glass bottle | |
1.3310 to 1.3330 (20°C, 589nm) | |
MFCD00006991 | |
01, 594 | |
0.785 | |
ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique | |
IKHGUXGNUITLKF-UHFFFAOYSA-N | |
acetaldehyde | |
177 | |
44.04 | |
Extra Pure |
Chemical Identifiers
75-07-0 | |
44.04 | |
IKHGUXGNUITLKF-UHFFFAOYSA-N | |
177 | |
acetaldehyde |
C2H4O | |
MFCD00006991 | |
ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique | |
CHEBI:15343 | |
CC=O |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Extremely flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
GHS Signal Word: Danger
EINECSNumber : 200-836-8
RUO – Research Use Only