Please login to your online account to display your discounted pricing

Alfa Aesar™ ACES potassium salt, 98%

$31.09 - $31.09

Chemical Identifiers

Chemical Name or Material ACES potassium salt
Molecular Formula H2NCOCH2NHCH2CH2SO3K
Formula Weight 220.29
MDL Number MFCD00065382
Synonym N-(2-Acetamido)-2-aminoethanesulfonic acid potassium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1549909

 
alfa aesar™
A1549909
10g Each for $31.09

AAA1549918

 
alfa aesar™
A1549918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1549930

 
alfa aesar™
A1549930
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 220.29
Chemical Name or Material ACES potassium salt
Assay 98%
Molecular Formula H2NCOCH2NHCH2CH2SO3K
MDL Number MFCD00065382
Synonym N-(2-Acetamido)-2-aminoethanesulfonic acid potassium salt
Quantity 10g