Please login to your online account to display your discounted pricing

ACES, >99%, Ultra Pure grade, MP Biomedicals™


Manufacturer: mp biomedicals  19397325

 View more versions of this product

Catalog No. ICN19397325

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material ACES
CAS 7365-82-4
Purity Grade UltraPure
Physical Form White crystalline powder
Assay Percent Range > 99%
Molecular Formula C4H10N2O4S
Formula Weight 182.2g/mol
Synonym 2-[(2-Amino-2-oxoethyl)amino]- ethanesulfonic acid

Specifications

Chemical Name or Material ACES
CAS 7365-82-4
Purity Grade UltraPure
Physical Form White crystalline powder
Assay Percent Range > 99%
Molecular Formula C4H10N2O4S
Formula Weight 182.2g/mol
Synonym 2-[(2-Amino-2-oxoethyl)amino]- ethanesulfonic acid