Please login to your online account to display your discounted pricing

ACES, >99%, Ultra Pure grade, MP Biomedicals™


Quantity 5g

Manufacturer: mp biomedicals  19397305

 View more versions of this product

Catalog No. ICN19397305

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 5g
Color White
Physical Form Crystalline Powder
Formula Weight 182.2
Purity Grade UltraPure
Chemical Name or Material ACES
Assay Percent Range > 99%
Molecular Formula C4H10N2O4S
CAS 7365-82-4
Synonym 2-[(2-Amino-2-oxoethyl)amino]- ethanesulfonic acid

Specifications

Chemical Name or Material ACES
CAS 7365-82-4
Purity Grade UltraPure
Physical Form Crystalline Powder
Assay Percent Range > 99%
Molecular Formula C4H10N2O4S
Formula Weight 182.2
Synonym 2-[(2-Amino-2-oxoethyl)amino]- ethanesulfonic acid