Please login to your online account to display your discounted pricing

ACES, >99%, Ultra Pure grade, MP Biomedicals™

$54.91 - $54.91

Chemical Identifiers

Chemical Name or Material ACES
CAS 7365-82-4
Purity Grade UltraPure
Physical Form Crystalline Powder
Assay Percent Range > 99%
Molecular Formula C4H10N2O4S
Formula Weight 182.2
Synonym 2-[(2-Amino-2-oxoethyl)amino]- ethanesulfonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19397305

 
mp biomedicals
19397305
5g Each for $54.91
Description & Specifications

Specifications

Quantity 5g
Color White
Physical Form Crystalline Powder
Formula Weight 182.2
Purity Grade UltraPure
Chemical Name or Material ACES
Assay Percent Range > 99%
Molecular Formula C4H10N2O4S
CAS 7365-82-4
Synonym 2-[(2-Amino-2-oxoethyl)amino]- ethanesulfonic acid