Please login to your online account to display your discounted pricing

ACES, >99%, MP Biomedicals™


Quantity 100g

Manufacturer: mp biomedicals  0210001180

 View more versions of this product

Catalog No. ICN10001180

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 100g
Chemical Name or Material Aces
CAS 7365-82-4
Physical Form White crystalline powder
Assay Percent Range >99%
Molecular Formula C4H10N2O4S
Formula Weight 182.2g/mol
MDL Number MFCD00008030
Synonym N-(2-Acetamido)-2-aminoethanesulfonic acid
EINECS Number 230-908-4

Specifications

Chemical Name or Material Aces
CAS 7365-82-4
Physical Form White crystalline powder
Assay Percent Range >99%
Molecular Formula C4H10N2O4S
Formula Weight 182.2g/mol
MDL Number MFCD00008030
Synonym N-(2-Acetamido)-2-aminoethanesulfonic acid