Please login to your online account to display your discounted pricing

ACES, >99%, MP Biomedicals™

$240.29 - $441.36

Chemical Identifiers

Chemical Name or Material Aces
CAS 7365-82-4
Physical Form White crystalline powder
Assay Percent Range >99%
Molecular Formula C4H10N2O4S
Formula Weight 182.2g/mol
MDL Number MFCD00008030
Synonym N-(2-Acetamido)-2-aminoethanesulfonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10001180

 
mp biomedicals
0210001180
100g Each for $240.29

ICN10001183

 
mp biomedicals
0210001183
250g Each for $441.36
Description & Specifications

Specifications

Quantity 100g
Chemical Name or Material Aces
CAS 7365-82-4
Physical Form White crystalline powder
Assay Percent Range >99%
Molecular Formula C4H10N2O4S
Formula Weight 182.2g/mol
MDL Number MFCD00008030
Synonym N-(2-Acetamido)-2-aminoethanesulfonic acid
EINECS Number 230-908-4