Please login to your online account to display your discounted pricing

ACES, 99%, extra pure, ACROS Organics™

$424.81 - $424.81

Chemical Identifiers

Chemical Name or Material ACES
Name Note Extra Pure
CAS 7365-82-4
CAS Min % 98.5
CAS Max % 100.0
Purity Grade Extra Pure
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C4H10N2O4S
Linear Formula H2NCOCH2NHCH2CH2SO3H
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC228002500

  SDS
thermo fisher scientific
AC228002500
Glass bottle 250g Each for $424.81

AC228001000

  SDS
fisher scientific acros
AC228001000
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC228000250

  SDS
fisher scientific acros
228000250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Extra Pure
Physical Form Crystalline Powder
Assay ≥98.5%
Formula Weight 182.19
Quantity 250g
Color White
Merck Index 15, 37
pH 3.6 to 4.4 (1% soln.)
Absorbance (c=5, H2O)
Absorbance Conditions (c=5, H2O)
Chemical Name or Material ACES
Name Note Extra Pure
Assay Percent Range 99%
Molecular Formula C4H10N2O4S
CAS 7365-82-4
Infrared Spectrum Authentic
Linear Formula H2NCOCH2NHCH2CH2SO3H
MDL Number MFCD00008030
CAS Min % 98.5
CAS Max % 100.0
Packaging Glass bottle
Synonym N-(2-Acetamido)-2-aminoethanesulfonic acid
Water 1% max. (K.F.)

Biological buffer