missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals ACES, 99+%, for biochemistry

ACES, >-99%, C4H10N2O4S, CAS Number-7365-82-4, N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine, 1kg, 0.02 max. at 280nm, 0.04 max. at 260nm, (5% aq. soln.), (5% aq. soln.) at 0nm, 99+%, 100.0, 99.0, CHEBI:39062, 100ppm max., White, 230-908-4, 182.19, Biochemical

Supplier:  Thermo Scientific Chemicals 327671000

 View more versions of this product

Catalog No. AC327671000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for biochemistry
Specifications

Specifications

(5% aq. soln.), (5% aq. soln.) at 0nm, 0.02 max. at 280nm, 0.04 max. at 260nm
(5% aq. soln.)
7365-82-4
100.0
3.6 to 4.4 (1% soln. at 25°C)
100 g
Authentic
H2NCOCH2NHCH2CH2SO3H
Plastic bottle
N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine
DBXNUXBLKRLWFA-UHFFFAOYSA-N
2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
81832
182.19
Biochemical
ACES
270.0°C to 295.0°C
99.0
White
Crystals
99+%
C4H10N2O4S
MFCD00008030
15, 37
Solubility in water: soluble (approx. 40g/L at 20 °C).
C(CS(=O)(=O)O)NCC(=O)N
182.194
CHEBI:39062
≥99%

Chemical Identifiers

7365-82-4
182.194
DBXNUXBLKRLWFA-UHFFFAOYSA-N
81832
2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
C4H10N2O4S
MFCD00008030
N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine
CHEBI:39062
C(CS(=O)(=O)O)NCC(=O)N
Safety and Handling

Safety and Handling

EINECSNumber : 230-908-4

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only