missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ ACES, 0.5M buffer soln., pH 7.5

$102.70 - $102.70

Chemical Identifiers

CAS 7365-82-4
Molecular Formula C4H10N2O4S
Molecular Weight (g/mol) 182.194
MDL Number MFCD00008030
InChI Key DBXNUXBLKRLWFA-UHFFFAOYSA-N
PubChem CID 81832
ChEBI CHEBI:39062
IUPAC Name 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
SMILES C(CS(=O)(=O)O)NCC(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61011AE View Documents Alfa Aesar™
J61011AE
100mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ61011AK View Documents Alfa Aesar™
J61011AK
250mL Each for $102.70
 
Specifications

Chemical Identifiers

7365-82-4
182.194
DBXNUXBLKRLWFA-UHFFFAOYSA-N
CHEBI:39062
C(CS(=O)(=O)O)NCC(=O)N
C4H10N2O4S
MFCD00008030
81832
2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid

Specifications

7365-82-4
MFCD00008030
DBXNUXBLKRLWFA-UHFFFAOYSA-N
2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
81832
Liquid
Colorless
ACES buffer soln.
C4H10N2O4S
0.5M buffer soln.
C(CS(=O)(=O)O)NCC(=O)N
182.194
CHEBI:39062
7.5
100mL
Safety and Handling

Safety and Handling

TSCA : TSCA

Recommended Storage : 2° to 8°C