missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ ACES, 0.5M buffer soln., pH 6.5

Manufacturer:  Alfa Aesar™ J60366AK

 View more versions of this product

Catalog No. AAJ60366AK



Specifications

Specifications

7365-82-4
MFCD00008030
DBXNUXBLKRLWFA-UHFFFAOYSA-N
2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
81832
Liquid
Colorless
C4H10N2O4S
0.5M buffer soln.
C(CS(=O)(=O)O)NCC(=O)N
182.194
CHEBI:39062
6.5
250mL

Chemical Identifiers

7365-82-4
182.194
DBXNUXBLKRLWFA-UHFFFAOYSA-N
CHEBI:39062
C(CS(=O)(=O)O)NCC(=O)N
C4H10N2O4S
MFCD00008030
81832
2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Keep cold

Documents