Learn More
Acenaphthenequinone, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2346614
Description
Acenaphthenequinone is used in the preparation of acetylcholinesterase inhibitory agents. It is also used in the preparation of turn-on sensors for cysteine/homocysteine and its application in bioimaging. Further, it is used as an intermediate in manufacturing of dyes and pharmaceuticals. In addition to this, it is used in chemical research as a drug and therapeutic agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAcenaphthenequinone is used in the preparation of acetylcholinesterase inhibitory agents. It is also used in the preparation of turn-on sensors for cysteine/homocysteine and its application in bioimaging. Further, it is used as an intermediate in manufacturing of dyes and pharmaceuticals. In addition to this, it is used in chemical research as a drug and therapeutic agent.
Solubility
Soluble in chloroform and alcohol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Acenaphthenequinone | |
253°C to 262°C | |
MFCD00003805 | |
879172 | |
Soluble in chloroform and alcohol. Insoluble in water. | |
C1=CC2=C3C(=C1)C(=O)C(=O)C3=CC=C2 | |
182.178 | |
CHEBI:15342 | |
95% |
82-86-0 | |
C12H6O2 | |
25 g | |
acenaphthenequinone, acenaphthoquinone, 1,2-acenaphthylenedione, acenaphthenedione, 1,2-acenaphthenequinone, 1,2-acenaphthenedione, acenaphthaquinone, acenaphthylene-1,2-quinone, acenaphthylenequinone, acenaphthodione | |
AFPRJLBZLPBTPZ-UHFFFAOYSA-N | |
acenaphthylene-1,2-dione | |
6724 | |
182.18 |
Chemical Identifiers
82-86-0 | |
182.178 | |
AFPRJLBZLPBTPZ-UHFFFAOYSA-N | |
6724 | |
acenaphthylene-1,2-dione |
C12H6O2 | |
MFCD00003805 | |
acenaphthenequinone, acenaphthoquinone, 1,2-acenaphthylenedione, acenaphthenedione, 1,2-acenaphthenequinone, 1,2-acenaphthenedione, acenaphthaquinone, acenaphthylene-1,2-quinone, acenaphthylenequinone, acenaphthodione | |
CHEBI:15342 | |
C1=CC2=C3C(=C1)C(=O)C(=O)C3=CC=C2 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 201-441-3
RTECSNumber : AB1024500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only