Learn More
Acenaphthene, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 201340050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Acenaphthene | |
98.5 | |
1.0600g/mL | |
135°C | |
98.5% min. (GC) | |
C12H10 | |
5g | |
1.06 | |
acenaphthene, 1,8-ethylenenaphthalene, naphthyleneethylene, peri-ethylenenaphthalene, acenaphthylene, 1,2-dihydro, ethylenenaphthalene, 1,8-dihydroacenaphthalene, unii-v8ut1gac5y, ccris 5951, v8ut1gac5y | |
CWRYPZZKDGJXCA-UHFFFAOYSA-N | |
1,2-dihydroacenaphthylene | |
6734 | |
154.21 |
83-32-9 | |
100.0 | |
279°C | |
Authentic | |
Glass Bottle | |
MFCD00003807 | |
05, IV, 1834 | |
15, 29 | |
Solubility in water: insoluble. Other solubilities: 3.2g/100mL glacial acetic acid,1g/31mL alcohol- 1g/56mL methanol,1g/25mL propanol- 1g/2.5mL chloroform,1g/5mL benzene- 1g/5mL toluene | |
C1CC2=C3C1=CC=CC3=CC=C2 | |
154.21 | |
CHEBI:22154 | |
99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning
EINECSNumber : 201-469-6
RUO â Research Use Only