Learn More
Acenaphthene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A149220B
Description
Acenaphthene is used in the preparation of naphthalene dicarboxylic anhydride, which is used as a precursor to dyes and optical brighteners. Further, it is used in the preparation of active pharmaceutical ingredients and in plastics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAcenaphthene is used in the preparation of naphthalene dicarboxylic anhydride, which is used as a precursor to dyes and optical brighteners. Further, it is used in the preparation of active pharmaceutical ingredients and in plastics.
Solubility
Soluble in chloroform, benzene, tolune, glacial acetic acid and methanol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and alkaline earth metals.
Specifications
Acenaphthene | |
92°C to 95°C | |
278°C | |
C12H10 | |
1000 g | |
386081 | |
acenaphthene, 1,8-ethylenenaphthalene, naphthyleneethylene, peri-ethylenenaphthalene, acenaphthylene, 1,2-dihydro, ethylenenaphthalene, 1,8-dihydroacenaphthalene, unii-v8ut1gac5y, ccris 5951, v8ut1gac5y | |
CWRYPZZKDGJXCA-UHFFFAOYSA-N | |
1,2-dihydroacenaphthylene | |
6734 | |
154.21 |
83-32-9 | |
1.15 | |
135°C (275°F) | |
MFCD00003807 | |
UN3077 | |
14,28 | |
Soluble in chloroform,benzene,tolune,glacial acetic acid and methanol. Insoluble in water. | |
C1CC2=C3C1=CC=CC3=CC=C2 | |
154.21 | |
CHEBI:22154 | |
97% |
Chemical Identifiers
83-32-9 | |
154.21 | |
CWRYPZZKDGJXCA-UHFFFAOYSA-N | |
6734 | |
1,2-dihydroacenaphthylene |
C12H10 | |
MFCD00003807 | |
acenaphthene, 1,8-ethylenenaphthalene, naphthyleneethylene, peri-ethylenenaphthalene, acenaphthylene, 1,2-dihydro, ethylenenaphthalene, 1,8-dihydroacenaphthalene, unii-v8ut1gac5y, ccris 5951, v8ut1gac5y | |
CHEBI:22154 | |
C1CC2=C3C1=CC=CC3=CC=C2 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 201-469-6
RTECSNumber : AB1000000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only