Please login to your online account to display your discounted pricing

ACDPP hydrochloride, Tocris

Selective mGlu5 receptor antagonist

$746.48 - $746.48

Chemical Identifiers

Chemical Name or Material ACDPP hydrochloride
Assay Percent Range >99%
Molecular Formula C12H13ClN6O·HCl
Formula Weight 329.19
Synonym 3-Amino-6-chloro-5-dimethylamino-N-2-pyridinylpyrazinecarboxamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

225450

 
r & d systems
RDS225450
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 329.19
Quantity 50mg
Chemical Name or Material ACDPP hydrochloride
Assay Percent Range >99%
Molecular Formula C12H13ClN6O·HCl
CAS 37804-11-8
Synonym 3-Amino-6-chloro-5-dimethylamino-N-2-pyridinylpyrazinecarboxamide hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature