Please login to your online account to display your discounted pricing

Thermo Scientific™ accuSpin™ 8C Benchtop Centrifuge Spacers


Accessories for accuSpin™ 8C small centrifuge

Manufacturer: thermo scientific™  75008817

 View more versions of this product

Catalog No. 13-101-506

  • / Pack of 8 Each
 

Description & Specifications

Specifications

For Use With (Equipment) accuSpin™ 8C Benchtop Centrifuge
Item Description Spacer
Color Green
Includes Set of 8
Length (Metric) 2cm