missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ accumet™ AB150 pH Meters

Intuitive, simple operation and high accuracy in a compact, affordable meter.

Manufacturer:  Fisherbrand™ ORI13636AB150P

Catalog No. 13-AB1-50P



Description

Description

  • Easy-to-understand prompts
  • Graphic display with adjustable backlight
  • Calibrate with custom pH buffer values
  • Date and Time to meet GLP
  • Password protection
  • Optional calibration reminder and high/low alarm
  • 500 point memory – log manually or at intervals
  • Easily view calibration data
  • 0.001 pH resolution and ±0.002 pH accuracy

pH Mode

  • Range: -2.000 to +20.000 pH
  • Resolution: 0.1/0.01/0.001 pH
  • Accuracy: ±0.002 pH + 1 LSD
  • Buffer Sets: USA: 2.000, 4.010, 6.997, 10.013, 12.000
  • NIST: 1.678, 4.010, 6.865, 9.184, 12.460
  • DIN: 1.090, 3.060, 4.650, 6.790, 9.230, 12.750
  • FSCI: 1.000, 3.000, 6.000, 8.000, 10.000, 13.000
  • CUSTOM: any two to five values, ≥1.0 pH unit apart
  • Slope Display: up to five different slopes with offset
  • Temp. Compensation: automatic or manual (0° to 100°C/32° to 212°F)
Output
  • RS-232 (phono plug), mini-B USB, stirrer
  • Language Selection
    • English, French, Spanish, German, Italian, Chinese, Korean, and Portuguese
  • Memory
    • 500 data sets, viewable
  • Datalogging
    • Manual, timed (selectable every 3 to 3600 seconds)
    • Printer or CSV format
    Cal Due Alarm
    • User selectable from 8 hours, 16 hours and 1 to 31 days
  • High/Low Alarms
    • User selectable, visual and audible, parameter based

    Specifications

    Specifications

    Electrode Arm, RS-232 & USB Cables, 100/240 V Power Supply, 13-620-631 pH/ATC Electrode
    32°F to 212°F
    0 to 14
    accumet AB150 pH Kit
    0°C to 100°C
    SDS
    Documents