Please login to your online account to display your discounted pricing
Accuform Signs Safety Projection Sign: Spill Response Equipment
Locate equipment or direct your way out with multi-angle view
$20.05 - $20.05
Specifications
For Use With (Application) For identification, location and directional purposes
Material Plastic
Message Spill Response Equipment
Type Projection sign
Quantity Each
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    
17-998-092
accuform mfg inc
PSP390
Blue, Black on White Each for $20.05
Description & Specifications
Specifications
For Use With (Application) For identification, location and directional purposes
Material Plastic
Message Spill Response Equipment
Type Projection sign
Quantity Each
Size (English) 6 x 5 in.
  • High-impact plastic with permanently bonded image on the two panels
  • Protected by over-laminate film
  • 1.5 in. side flanged edges have pre-drilled mounting holes - includes screws and self-adhesive foam tape
  • 90D signs project at 90° from wall; measure 8 x 8 in.
  • 3D signs project at 45° from either side; measure 6 x 5 in.
  • Signs made of Lumi-Glow™ material glow in the dark
Screws and self-adhesive foam tape