missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Accuform Signs Safety Projection Sign: First Aid

When you need identification, location, and direction, signs can be seen from various angles for extra visibility.

$21.14 - $21.14

Specifications

Color Green/Black/White
For Use With (Application) Call attention to the location of first aid equipment that is placed throughout the facility
Message First Aid
Quantity 1 Ea.
Shape V-shaped
View More Specs

Products 1
Catalog Number Mfr. No. Style Price Quantity  
Catalog Number Mfr. No. Style Price Quantity  
17-998-354
View Documents
Accuform Signs
PSP605
3D
Each for $21.14
 
Description

Description

  • Hang signs along hallways, stairways, corridors and doorways
  • Signs are .10 in. thick high-impact plastic
  • Images on the two panels are protected by an over-laminate
  • Flanged edges have pre-drilled mounting holes
  • Wall mount with screws or self-adhesive foam tape (both included)

Available in two styles:

  • 3D signs project 45 degrees from the wall, providing optimum viewing from the front and sides
  • 90D signs project 90 degrees from the wall and can be viewed from either side

Specifications

Specifications

Green/Black/White
First Aid
V-shaped
Projection sign
Call attention to the location of first aid equipment that is placed throughout the facility
1 Ea.
10 in.
Plastic
SDS