Please login to your online account to display your discounted pricing

Accuform Signs Caution Watch Your Step Sign

Colorful, compact safety signs

$20.15 - $20.15

Specifications

Certifications/Compliance ANSI Z535.2-22, 29 CFR 191.145
Color Black on Yellow
Dimensions (L x W) 10 x 14 in. (25.4 x 35.6cm)
For Use With (Application) Intended to indicate potential, type and degree of the hazard that may lead to accidental injury or property damage
Height (English) 10 in.
View More Specs
Products
Catalog Number Mfr. No. Material Temperature Range (English) Temperature Range (Metric) Thickness Price Quantity    

19-804-704

 
accuform mfg inc
MCSPC37VP
Plastic -90 to 168°F -68 to 76°C 0.06 in. Each for $20.15
Description & Specifications

Specifications

Certifications/Compliance ANSI Z535.2-22, 29 CFR 191.145
Color Black on Yellow
Dimensions (L x W) 10 x 14 in. (25.4 x 35.6cm)
For Use With (Application) Intended to indicate potential, type and degree of the hazard that may lead to accidental injury or property damage
Height (English) 10 in.
Message Watch Your Step
Type Safety sign
Languages English
Length (English) 10 in.
Length (Metric) 25.4cm
Quantity Each
Style Surface Mount
Width (English) 14 in.
Width (Metric) 35.6cm

  • Legend: CAUTION WATCH YOUR STEP
  • Size: 25.4 × 35.6cm (10 × 14 in.)
  • Color: Yellow, Black
  • Language: English