Please login to your online account to display your discounted pricing
Thermo Scientific™ Accucore™ Phenyl-Hexyl LC Columns
Offer unique selectivity for aromatic and moderately polar analytes with Accucore Phenyl-Hexyl LC Columns.
$774.89 - $1,708.56
Specifications
Carbon Load 5%
Column Type Reversed Phase
Particle Shape Spherical Solid Core
Particle Size 2.6µm
Pore Size 80Å
View More Specs
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products
Catalog Number Mfr. No. Column Format Diameter (Metric) Length (Metric) Particle Size Price Quantity    
17-926-052130
thermo scientific™
17926052130
Analytical Column 2.1mm 50mm 2.6µm Each for $1,100.72
17-926-053030
thermo scientific™
17926053030
Analytical Column 3mm 50mm 2.6µm Each for $1,263.09
17-926-054630
thermo scientific™
17926054630
Analytical Column 4.6mm 50mm 2.6µm Each for $1,263.09
17-926-102130
thermo scientific™
17926102130
Analytical Column 2.1mm 100mm 2.6µm Each for $1,314.00
17-926-103030
thermo scientific™
17926103030
Analytical Column 3mm 100mm 2.6µm Each for $1,297.72
17-926-104630
thermo scientific™
17926104630
Analytical Column 4.6mm 100mm 2.6µm Each for $1,484.83
17-926-152130
thermo scientific™
17926152130
Analytical Column 2.1mm 150mm 2.6µm Each for $1,708.56
17-926-153030
thermo scientific™
17926153030
Analytical Column 3mm 150mm 2.6µm Each for $1,708.56
17-926-154630
thermo scientific™
17926154630
Analytical Column 4.6mm 150mm 2.6µm Each for $1,708.56
17-926-013005
thermo scientific™
17926013005
Guard Cartridge, 4 PK 3mm 10mm 2.6µm Pack of 4 Each for $774.89
17-926-012105
thermo scientific
17926012105
Guard Cartridge, 4 PK 2.1mm 10mm 2.6µm Pack of 4 Each for $788.40
17-926-014005
thermo scientific™
17926014005
Guard Cartridge, 4 PK 4mm 10mm 2.6µm Pack of 4 Each for $886.61
17-926-032130
thermo scientific
17926032130
Analytical Column 2.1mm 30mm 2.6µm Each for $1,039.39
Description & Specifications
Specifications
Carbon Load 5%
Column Type Reversed Phase
Particle Shape Spherical Solid Core
Particle Size 2.6µm
Pore Size 80Å
Endcapped Yes
Max. Pressure 1000bar
Packing Material Spherical, solid core ultrapure silica
pH 2 to 8
Surface Area 130m²/g
Temperature 70°C
USP Type L11
Offer unique selectivity for aromatic and moderately polar analytes with Thermo Scientific™ Accucore™ Phenyl-Hexyl LC Columns.
Based on Core Enhanced Technology, Accucore columns provide fast, high-resolution separations without the elevated backpressures required by sub-2μm particles.
  • Fast separations with excellent resolution and sensitivity
  • Lower backpressures so that UHPLC is not required
  • Mixed-Mode Selectivity for Aromatic and Moderately Polar Analytes
  • Enhanced Pi-pi Interactions with Aromatics
  • Moderate Hydrophobicity
Ordering Information:
Other custom column dimensions are available. Please call your local Customer Service for more information.