Please login to your online account to display your discounted pricing

Thermo Scientific™ Accucore™ 150-C4 LC Columns


Lower hydrophobic retention, ideal for optimal retention of proteins and larger peptides.

Manufacturer: thermo scientific™  16526053030

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. 03-898-121

  • / Each
 

Description & Specifications

Specifications

Diameter (Metric) 3mm
Column Format Analytical Column
Length (Metric) 50mm
Surface Area 80m²/g
USP Type L26
Column Type Reversed Phase
Carbon Load 2%
Packing Material Spherical, solid core ultrapure silica
Max. Pressure 1000bar
Temperature 60°C
pH 2 to 9
Endcapped Yes
For Use With HPLC
Particle Size 2.6µm
Pore Size 150Å
Stationary Phase C4

Offer lower hydrophobic retention, ideal for optimal retention of proteins and larger peptides with Thermo Scientific™ Accucore™ 150-C4 LC Columns. Based on Core Enhanced Technology, Thermo Scientific Accucore columns provide fast, high-resolution biomolecule separations without the elevated backpressures required by sub-2μm particles.

  • 150Å pore size solid core particles for fast biomoleucule separations
  • Superb resolution at low backpressures
  • Exceptionally rugged columns
  • Significantly lower hydrophobic retention than C18
  • Ideal for optimal retention of proteins and larger peptides

Ordering Information:

Other custom column dimensions are available. Please call your local Customer Service for more information.