Thermo Scientific™ Accucore™ 150-C4 LC Columns
Lower hydrophobic retention, ideal for optimal retention of proteins and larger peptides.
Manufacturer:  Thermo Scientific™ 16526013005
 View more versions of this product
Catalog No. 03-898-130
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Offer lower hydrophobic retention, ideal for optimal retention of proteins and larger peptides with Thermo Scientific™ Accucore™ 150-C4 LC Columns. Based on Core Enhanced Technology, Thermo Scientific Accucore columns provide fast, high-resolution biomolecule separations without the elevated backpressures required by sub-2μm particles.
  • 150Å pore size solid core particles for fast biomoleucule separations
  • Superb resolution at low backpressures
  • Exceptionally rugged columns
  • Significantly lower hydrophobic retention than C18
  • Ideal for optimal retention of proteins and larger peptides
Ordering Information:
Other custom column dimensions are available. Please call your local Customer Service for more information.
Description & Specifications
Specifications
2%
Reversed Phase
Yes
10mm
Silica, Spherical Solid Core Ultrapure
2 to 9
80m²/g
L26
Guard Cartridge, 4 PK
3mm
HPLC
1000bar
2.6μm
150Å
60°C
C4