Please login to your online account to display your discounted pricing

Thermo Scientific Accucore™ 150 C18 LC Columns


Offer phase characteristics designed for the separation of peptides with Thermo Scientific™ Accucore™ 150 C18 LC Columns.

Manufacturer: thermo scientific  16126032130

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. 03-898-100

  • / Each
 

Description & Specifications

Specifications

Particle Size 2.6µm
Stationary Phase C18
pH 1 to 11
Pore Size 150Å
Temperature 60°C
Length (Metric) 30mm
Diameter (Metric) 2.1mm
Column Format Analytical Column
Max. Pressure 1000bar
For Use With HPLC
Surface Area 80m²/g
Column Type Reversed Phase
Packing Material Spherical, solid core ultrapure silica
USP Type L1
Carbon Load 7%
Endcapped Yes

Offer phase characteristics designed for the separation of peptides with Thermo Scientific™ Accucore™ 150 C18 LC Columns. Based on Core Enhanced Technology, Accucore columns provide fast, high-resolution biomolecule separations without the elevated backpressures required by sub-2μm particles.

  • 150Å pore size solid core particles for fast biomoleucule separations
  • Superb resolution at low backpressures
  • Exceptionally rugged columns
  • Designed for the separation of peptides Outstanding resolution

Ordering Information:

Other custom column dimensions are available. Please call your local Customer Service for more information.