missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bioscience accu-Test™ Vials for COD Testing
For low-, standard- and high-range concentrations
Manufacturer: Bioscience 185252
Includes: 16mm vial with 1.5mL digestion solution, 3.5mL catalyst solution, and screw cap
Description
- Convenient to use
- Give excellent analytical results
- Compatible with all standard COD reactors and spectrophotometers
- For samples with low chloride and ammonia levels, mercury-free reagents are available for nonreporting purposes
Order Info
Supplied in modular packs that are easy to stack
Certifications
U.S. EPA approved (for standard and low ranges) micro Chemical Oxygen Demand test
Recommended Storage
Shelf Life: one year from date of manufacture
Specifications
EPA, OSHA | |
Mercury free standard range concentration | |
Standard COD reactor and spectrophotometer |
75% Sulfuric Acid, 0.20% Potassium Dichromate, 0.40% Silver Sulfate, 0.002%Sulfamic Acid | |
COD testing for 20 to 900mg/L standard range concentration | |
16mm vial with 1.5mL digestion solution, 3.5mL catalyst solution, and screw cap |