Please login to your online account to display your discounted pricing
Thermo Scientific™ Acclaim™ SST Guard Cartridge Holders
Thermo Scientific™ Acclaim SST Guard Cartridge Holders are version 1 and 2 for seemless connection with Acclaim columns
$364.14 - $364.14
Specifications
For Use With Guard Cartridge
Type Guard Cartridge Holder
View More Specs
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products
Catalog Number Mfr. No. Item Description Price Quantity    
DX069580
thermo scientific™
069580
V-2 Each for $364.14
Description & Specifications
Specifications
For Use With Guard Cartridge
Type Guard Cartridge Holder
Version 1 and 2 for seemless connection with Acclaim columns.