Please login to your online account to display your discounted pricing

Thermo Scientific™ PepMap™ 100 C8 LC Columns

For high-resolution separation of very hydrophobic peptides, use this Thermo Scientific™ PepMap™ 100 C8 LC Columns as an excellent alternative to the PepMap C18 column.

$721.45 - $1,250.49

Specifications

Carbon Load 9%
Column Type Reversed Phase
Endcapped Yes
Material Stainless Steel
Max. Pressure 500bar
View More Specs
Products
Catalog Number Mfr. No. Column Format Diameter (Metric) Length (Metric) Particle Size Price Quantity    

DX161185

 
thermo scientific™
161185
Analytical Column 0.075mm 150mm 3µm Each for $1,250.49

DX161194

 
thermo scientific™
161194
Trap Cartridge 0.3mm 5mm 5µm Pack of 5 Each for $721.45
Description & Specifications

Specifications

Carbon Load 9%
Column Type Reversed Phase
Endcapped Yes
Material Stainless Steel
Max. Pressure 500bar
Packing Material Spherical, fully porous ultrapure silica
pH 2 to 8
Pore Size 100Å
Stationary Phase C8
Surface Area 450m²/g
Temperature 60°C
USP Type L7

Choose Thermo Scientific™ Acclaim™ PepMap™ 100 C8 LC Columns when separating very hydrophobic peptides (nontryptic peptides) as an alternative to the PepMap™ C18 column.

This column provides high-resolution, high-sensitivity protein identification for proteomics and biomarker discovery and verification, and is ideal for:

 

High loading capcity

Acclaim PepMap columns are exceptionally suitable for the analysis of low abundant peptides often found in complex proteomics samples.

The PepMap 100 C8 LC Columns is available in 150mm length with a choice of 75, 300, 1000, and classic 75µm I.D. to allow tuning for throughput, sensitivity and amount of sample available.

Superior protein and peptide separations

Trifluoroacetic acid (TFA)-free LC/MS minimizes suppression effects for enhanced MS sensitivity

Outstanding separation efficiency

Superior resolution and recovery

Easy installation with Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fittings

Columns are available with nanoViper Fingertight Fittings

Requires no tools

Provides virtually zero-dead-volume connections

Withstands pressures up to a 1000 bar

About Nano, capillary, and micro Acclaim PepMap columns

Nano (75µm), capillary (300µm) and micro (1000µm) columns are ideal for protein and peptide analyses. Sample amount is often limited and smaller I.D. columns can increase in sensitivity in sample-limited situations. While nano LC columns are often used in proteomics, the concentrating effects of capillary and micro columns are beneficial for metabolomics or for facilitating LC-MS interfacing. Thermo Scientific Acclaim PepMap columns are offered in nano, capillary and micro column in a variety of chemistries for peptides and protein analysis.