Please login to your online account to display your discounted pricing
Thermo Scientific™ Titanium-Dioxide Nano-Trap Columns
Titanium-Dioxide Nano-Trap Columns support the enrichment of phosphopeptides.
$382.37 - $485.71
Specifications
Carbon Load -
Column Format Trap Column
Column Type Peptide
Endcapped -
Max. Pressure 500bar
View More Specs
Products
Catalog Number Mfr. No. Column Format Diameter (Metric) Length (Metric) Particle Size Price Quantity    
DX164215
thermo scientific™
164215
Trap Column 0.2mm 10mm 5µm Pack of 2 Each for $382.37
DX164205
thermo scientific™
164205
Trap Column 0.1mm 10mm 5µm Pack of 2 Each for $485.71
Description & Specifications
Specifications
Carbon Load -
Column Format Trap Column
Column Type Peptide
Endcapped -
Max. Pressure 500bar
pH -
Pore Size -
Stationary Phase Acclaim Titanium dioxide
Surface Area -
Temperature -
Type Trap Column
USP Type N/A
Support the enrichment of phosphopeptides with Thermo Scientific™ Titanium-Dioxide Nano-Trap Columns.