Please login to your online account to display your discounted pricing

Thermo Scientific™ Titanium-Dioxide Nano-Trap Columns


Titanium-Dioxide Nano-Trap Columns support the enrichment of phosphopeptides.

Manufacturer: thermo scientific™  164205

 View more versions of this product

Catalog No. DX164205

  • / Pack of 2 Each
 

Description & Specifications

Specifications

Column Format Trap Column
Diameter (Metric) 0.1mm
pH -
Length (Metric) 10mm
Pore Size -
Packing Material Titanium dioxide
Type Trap Column
Temperature -
Particle Size 5µm
Surface Area -
Stationary Phase TiO2
Material Fused Silica
USP Type N/A
Max. Pressure 500bar
Endcapped -
Carbon Load -
Column Type Reversed Phase

Support the enrichment of phosphopeptides with Thermo Scientific™ Titanium-Dioxide Nano-Trap Columns.