Please login to your online account to display your discounted pricing

Thermo Scientific™ Titanium-Dioxide Nano-Trap Columns


Titanium-Dioxide Nano-Trap Columns support the enrichment of phosphopeptides.

Manufacturer: thermo scientific™  164215

 View more versions of this product

Catalog No. DX164215

  • / Pack of 2 Each
 

Description & Specifications

Specifications

Column Format Trap Column
Diameter (Metric) 0.2mm
Stationary Phase TiO2
Column Type Reversed Phase
Temperature -
Surface Area -
Carbon Load -
Type Trap Column
Endcapped -
Particle Size 5µm
Max. Pressure 500bar
Length (Metric) 10mm
USP Type N/A
pH -
Packing Material Titanium dioxide
Pore Size -
Material Fused Silica

Support the enrichment of phosphopeptides with Thermo Scientific™ Titanium-Dioxide Nano-Trap Columns.