Please login to your online account to display your discounted pricing

Thermo Scientific™ PepMap™ 300 C4 LC Columns


Separate hydrophobic peptides and proteins, providing higher recoveries with Thermo Scientific™ PepMap™ 300 C4 LC Columns.

Manufacturer: thermo scientific™  163591

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. DX163591

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Column Format Trap Cartridge
Diameter (Metric) 0.3mm
pH 2 to 8
Pore Size 300Å
Type Trap Cartridge
Temperature 60°C
Particle Size 5µm
Column Type Reversed Phase
Length (Metric) 5mm
Max. Pressure 500bar
Surface Area 450m²/g
USP Type L26
Endcapped Yes
Carbon Load 3%
For Use With LC/MS, ESI-MS, MALDI-MS
Stationary Phase C4

Separate hydrophobic peptides and proteins, providing higher recoveries with Thermo Scientific™ Acclaim™ PepMap™ 300 C4 LC Columns.

  • High resolution in protein identification, biomarker discovery and systems biology
  • Particular loadability allows hghest sensitivity in LC/MS
  • Designed for TFA-free LC-MS, minimizing ion-suppression effects
  • Optimized for coupling to ESI/MS and MALDI-MS
  • Highest column-to-column reproducibility
  • Easy-to-use, cutting-edge miniaturized HPLC