Please login to your online account to display your discounted pricing

Thermo Scientific™ Acclaim™ 5μm PepMap™ 300 μ-Precolumns™ Cartridge Columns


Thermo Scientific™ Acclaim™ 5μm PepMap™ 300 μ-Precolumns Cartridge Columns very short microcolumns consisting of a cartridge holder and a set of disposable cartridges.

Manufacturer: thermo scientific™  163593

 View more versions of this product

Catalog No. DX163593

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Column Type Peptide
Particle Size 5µm
Pore Size 300Å
For Use With LC/MS, ESI-MS, MALDI-MS
Stationary Phase Acclaim PepMap C18
Length (Metric) 15mm
Material Stainless Steel
Diameter (Metric) 1mm

Thermo Scientific™ Acclaim™ 5μm PepMap™ 300 μ-Precolumns™ Cartridge Columns are very short microcolumns consisting of a cartridge holder and a set of disposable cartridges.

  • For sample pre-concentration, sample cleanup, or in various column-switching techniques
  • 300Å pore size