Please login to your online account to display your discounted pricing

Thermo Scientific™ Acclaim™ 300 C18 LC Columns


Achieve high-resolution peptide mapping and protein separations with reversed-phase Thermo Scientific™ Acclaim™ 300 C18 LC Columns.

Manufacturer: thermo scientific™  069690

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. DX069690

  • / Pack of 2 Each
 

Description & Specifications

Specifications

Material
Packing Material
Flow Rate 1mL/min
Column Format Guard Cartridge, 2 PK
Diameter (Metric) 2.1mm
Length (Metric) 10mm
Surface Area 100m²/g
Carbon Load 8%
For Use With LC/MS
Column Type Reversed Phase
Type Guard Cartridge, 2PK
Stationary Phase C18
Endcapped Yes
Max. Pressure 6000psi
Particle Size 5µm
USP Type L1
Temperature 60°C
pH 2.5 to 7.5
Pore Size 300Å

Achieve high-resolution peptide mapping and protein separations with Thermo Scientific™ Acclaim™ 300 C18 LC Columns. The 3µm ultrapure silica particles reduce the limits for diffusion of the analytes into the stationary phase, resulting in fast, high-resolution separations. Compared to 5µm column packings, the smaller particles support increased flow rates and shallower gradients on shorter columns for faster separations.

High-Resolution Separation of Proteins and Peptides

  • High-performance C18 bonding chemistry on 300Å pore silica
  • Application tested for suitability in peptide mapping
  • Reproducible for dependable results
  • Extremely low bleed, fully compatible with MS
  • Stable between pH 2 .5 and 7.5
About the Acclaim Family of HPLC Columns
  • Complementary selectivity, high column efficiencies and ultrahigh resolution
  • Broad spectrum of stationary phases, including conventional C8 and C18 phases, polar embedded phases and specialty phases including wide-pore
  • Performance tested to ensure high quality and reliability