Please login to your online account to display your discounted pricing

Thermo Scientific™ Acclaim™ 300 C18 LC Columns

Achieve high-resolution peptide mapping and protein separations with reversed-phase Thermo Scientific™ Acclaim™ 300 C18 LC Columns.

$277.72 - $1,158.97

Specifications

Carbon Load 8%
Column Type Reversed Phase
Endcapped Yes
For Use With LC/MS
pH 2.5 to 7.5
View More Specs

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Products
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Particle Size Price Quantity    

DX060264

 
thermo scientific™
060264
2.1mm 150mm 3µm Each for $1,158.97

DX060266

 
thermo scientific™
060266
4.6mm 150mm 3µm Each for $1,158.97

DX063684

 
thermo scientific™
063684
3mm 150mm 3µm Each for $1,158.97

DX069690

 
thermo scientific™
069690
2.1mm 10mm 5µm Pack of 2 Each for $333.26

DX069697

 
thermo scientific™
069697
4.6mm 10mm 5µm Pack of 2 Each for $277.72

DX060263

 
thermo scientific™
060263
2.1mm 50mm 3µm Each for $765.37

DX060265

 
thermo scientific™
060265
4.6mm 50mm 3µm Each for $765.37
Description & Specifications

Specifications

Carbon Load 8%
Column Type Reversed Phase
Endcapped Yes
For Use With LC/MS
pH 2.5 to 7.5
Pore Size 300Å
Stationary Phase C18
Surface Area 100m²/g
Temperature 60°C
USP Type L1

Achieve high-resolution peptide mapping and protein separations with Thermo Scientific™ Acclaim™ 300 C18 LC Columns. The 3µm ultrapure silica particles reduce the limits for diffusion of the analytes into the stationary phase, resulting in fast, high-resolution separations. Compared to 5µm column packings, the smaller particles support increased flow rates and shallower gradients on shorter columns for faster separations.

High-Resolution Separation of Proteins and Peptides

  • High-performance C18 bonding chemistry on 300Å pore silica
  • Application tested for suitability in peptide mapping
  • Reproducible for dependable results
  • Extremely low bleed, fully compatible with MS
  • Stable between pH 2 .5 and 7.5
About the Acclaim Family of HPLC Columns
  • Complementary selectivity, high column efficiencies and ultrahigh resolution
  • Broad spectrum of stationary phases, including conventional C8 and C18 phases, polar embedded phases and specialty phases including wide-pore
  • Performance tested to ensure high quality and reliability