Please login to your online account to display your discounted pricing

Accessories for Sper Scientific Meters

Intelligent probes electronically retain calibration information.

$134.71 - $208.57

Specifications

For Use With (Application) Water quality testing
View More Specs
Products
Catalog Number Mfr. No. For Use With (Equipment) Product Type Price Quantity    

S96277

 
sper scientific
840016
Sper Scientific meters pH Probe Each for $208.57

S96439

 
sper scientific
840038
pH meter 840035 ATC Replacement Probe Each for $134.71

S96291

 
sper scientific
850059P
Sper Scientific meters ATC pH Probe, for low ionic Water Each for $136.07
Description & Specifications

Specifications

For Use With (Application) Water quality testing

  • Uses interchangeable probes to read pH, mV, conductivity, TDS, DO temperature and ORP