Please login to your online account to display your discounted pricing
American Educational Products Accessories for Jewel Oceanic Systems
Enhance your Jewel Oceanic system with these accessories
$1,373.71 - $1,373.71
Specifications
For Use With (Equipment) Jewel Oceanic 55 gal. system
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Material Product Type Price Quantity    
S17964AMF
hubbard scientific co
35645EMW
Create a completely free standing unit Iron Stationary Stand Each for $1,373.71
Description & Specifications
Specifications
For Use With (Equipment) Jewel Oceanic 55 gal. system
Light strip
  • Thermoplastic fixture with fluorescent tube
  • Tube fits on glass cover to provide supplemental illumination inside the tank
Stand
  • 30 in. high
  • Equipped with 4 in. ball-bearing casters with lockable brakes
Hydrometer/thermometer
  • Replacement for hydrometer/thermometer included with the Jewel Oceanic