Fisherbrand™ Accessories for Tube Rotator
14- to 63-place carousels for 11 to 34mm tubes
Manufacturer:  Fisherbrand™ BKL260651
 View more versions of this product
Catalog No. 05-450-202
Description
Description
Tube Rotator is Ideal for Biochemistry, Molecular Biology and Clinical applications. 14- to 63-place carousels for 11 to 34mm tubes
Specifications
Specifications
46 x 9-11mm
1.5/2mL
Fisher Scientific tube rotator
Carousel
9 to 11mm
46.0
Documents