missing translation for 'onlineSavingsMsg'
Learn More

Acamprosate Calcium 98.0+%, TCI America™

Supplier:  TCI America A23591G

Catalog No. A23591G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Acamprosate Calcium
270°C
C10H20CaN2O8S2
1 g
BUVGWDNTAWHSKI-UHFFFAOYSA-L
calcium;3-acetamidopropane-1-sulfonate
155434
400.47
Crystalline Powder
77337-73-6
White
MFCD00886588
Calcium 3-Acetamido-1-propanesulfonate, 3-Acetamido-1-propanesulfonic Acid Calcium Salt
CC(=O)NCCCS(=O)(=O)[O-].CC(=O)NCCCS(=O)(=O)[O-].[Ca+2]
400.474
CHEBI:51042
≥98.0% (HPLC,N)

Chemical Identifiers

77337-73-6
400.474
BUVGWDNTAWHSKI-UHFFFAOYSA-L
155434
calcium;3-acetamidopropane-1-sulfonate
C10H20CaN2O8S2
MFCD00886588
Calcium 3-Acetamido-1-propanesulfonate, 3-Acetamido-1-propanesulfonic Acid Calcium Salt
CHEBI:51042
CC(=O)NCCCS(=O)(=O)[O-].CC(=O)NCCCS(=O)(=O)[O-].[Ca+2]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : TZ6357000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions