Please login to your online account to display your discounted pricing

(±)-AC 7954 hydrochloride, Tocris

Non-peptide UT receptor agonist

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material (±)-AC 7954 hydrochloride
Assay Percent Range >98%
Molecular Formula C19H20ClNO2·HCl
Formula Weight 366.28
Synonym (±)-3-(4-Chlorophenyl)-3-[2-(dimethylamino)ethyl]-3, 4-dihydro-1H-2-benzopyran-1-one hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

248410

 
r & d systems
RDS248410
10mg Each for $244.81

248450

 
r & d systems
RDS248450
50mg Each for $997.23
Description & Specifications

Specifications

Formula Weight 366.28
Quantity 10mg
Chemical Name or Material (±)-AC 7954 hydrochloride
Assay Percent Range >98%
Synonym (±)-3-(4-Chlorophenyl)-3-[2-(dimethylamino)ethyl]-3, 4-dihydro-1H-2-benzopyran-1-one hydrochloride
Molecular Formula C19H20ClNO2·HCl
CAS 477313-09-0
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C