missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

AC 55649, Tocris

Selective RARβ2 agonist

$166.60 - $693.49

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C21H26O2
Formula Weight 310.44
Synonym 4'-Octyl-[1, 1'-biphenyl]-4-carboxylic acid
Chemical Name or Material AC 55649
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
243610 R & D Systems
RDS243610
10mg Each for $166.60
243650 R & D Systems
RDS243650
50mg Each for $693.49
Specifications Description & Specifications

Chemical Identifiers

>99%
310.44
AC 55649
C21H26O2
4'-Octyl-[1, 1'-biphenyl]-4-carboxylic acid

Specifications

59662-49-6
C21H26O2
4'-Octyl-[1, 1'-biphenyl]-4-carboxylic acid
AC 55649
>99%
310.44
10mg

Safety and Handling

Recommended Storage : Store at +4°C