Please login to your online account to display your discounted pricing

AC 55541, Tocris

Potent and selective PAR2 agonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material AC 55541
Assay Percent Range >97%
Molecular Formula C25H20BrN5O3
Formula Weight 518.36
Synonym (2E)-2-[1-(3-Bromophenyl)ethylidene] α-(benzoylamino)-3, 4-dihydro-4-oxo-1-phthalazineacetic acid hydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

336910

 
r & d systems
336910
10mg Each for $252.39

336950

 
r & d systems
336950
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 518.36
Quantity 10mg
Assay Percent Range >97%
CAS 916170-19-9
Chemical Name or Material AC 55541
Synonym (2E)-2-[1-(3-Bromophenyl)ethylidene] α-(benzoylamino)-3, 4-dihydro-4-oxo-1-phthalazineacetic acid hydrazide
Molecular Formula C25H20BrN5O3
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C