Please login to your online account to display your discounted pricing

AC 45594, Tocris

SF-1 inverse agonist

$136.87 - $580.08

Chemical Identifiers

Chemical Name or Material AC 45594
Assay Percent Range >99%
Molecular Formula C13H20O2
Formula Weight 208.3
Synonym 4-(Heptyloxy)phenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

304310

 
r & d systems
RDS304310
10mg Each for $136.87

304350

 
r & d systems
RDS304350
50mg Each for $580.08
Description & Specifications

Specifications

Formula Weight 208.3
Quantity 10mg
Chemical Name or Material AC 45594
Assay Percent Range >99%
Molecular Formula C13H20O2
Synonym 4-(Heptyloxy)phenol
Recommended Storage Store at +4°C
CAS 13037-86-0
Safety and Handling

Store at +4°C