Please login to your online account to display your discounted pricing

AC 264613, Tocris

PAR2 receptor agonist

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material AC 264613
Assay Percent Range >98%
Molecular Formula C19H18BrN3O2
Formula Weight 400.27
Synonym (±)-(3R™, 4S™)-2-Oxo-4-phenyl-3-pyrollidinecarboxylic acid 2-[1-(3-bromophenyl)ethylidene]hydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

337010

 
r & d systems
RDS337010
10mg Each for $218.74

337050

 
r & d systems
RDS337050
50mg Each for $915.10
Description & Specifications

Specifications

Formula Weight 400.27
Quantity 10mg
Assay Percent Range >98%
Molecular Formula C19H18BrN3O2
Chemical Name or Material AC 264613
CAS 1051487-82-1
Synonym (±)-(3R™, 4S™)-2-Oxo-4-phenyl-3-pyrollidinecarboxylic acid 2-[1-(3-bromophenyl)ethylidene]hydrazide
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C