Please login to your online account to display your discounted pricing

ABT 751, Tocris™

Inhibitor of microtubule polymerization; antimitotic and antitumor

$270.88 - $1,140.36

Chemical Identifiers

Chemical Name or Material ABT 751
Assay Percent Range >98%
Molecular Formula C18H17N3O4S
Formula Weight 371.41
Synonym N-[2-[(4-Hydroxyphenyl)amino]-3-pyridinyl]-4-methoxybenzenesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

413850

 
r & d systems
RDS413850
50mg Each for $1,140.36

413810

 
r & d systems
RDS413810
10mg Each for $270.88
Description & Specifications

Specifications

Formula Weight 371.41
Quantity 50mg
Molecular Formula C18H17N3O4S
CAS 141430-65-1
Recommended Storage Store at +4°C
Chemical Name or Material ABT 751
Assay Percent Range >98%
Synonym N-[2-[(4-Hydroxyphenyl)amino]-3-pyridinyl]-4-methoxybenzenesulfonamide
Safety and Handling

Store at +4°C