missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ ABsolute™ Probe-Based Detection qPCR Master Mixes

Thermo Scientific™ ABsolute qPCR Master Mixes offer sensitivity and reproducibility for real-time PCR applications.

Manufacturer:  Thermo Scientific™ AB1132B

 View more versions of this product

Catalog No. AB-1132B


 

Disclaimers:

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.


Specifications

Description

Enjoy sensitivity and reproducibility in real-time PCR applications with Thermo Scientific™ ABsolute™ Probe-Based Detection qPCR Master Mixes.

  • High sensitivity & specificity - The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
  • Reproducibility - All mixes undergo extensive QC to ensure batch to batch consistency
  • Optimized buffer - For maximal performance in multiplex reactions
Description & Specifications

Specifications

heated to 95°C for 15 minutes
Detection Method: Primer-Probe Detection, PCR Method: qPCR
Bio-Rad CFX384 Touch™, BioRad CFX96™, BioRad MyiQ, BioRad iCycler iQ, BioRad iQ5, Cepheid SmartCycler, Corbett Rotor-Gene, Eppendorf MasterCycler RealPlex, MJ Chromo4, MJ Opticon, Roche LightCycler 480
1600 Reactions
DNA, dsDNA
ABsolute QPCR Mix, QPCR mix; 1.25mL vial; 800 rxns
Real time PCR (qPCR), gene expression analysis, siRNA validation, genotyping
16 x 1.25mL ABsolute QPCR Mix (2X)
16 x 1.25mL vials; 800 x 50µL rxns
Store at -20°C.